siRNA Details
| Gene Symbol: Macc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163136.1 | NP_001156608.1 | 4 | 3015 | ||
| GenBank mRNA | AK028965.1 | - | 3 | 1337 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Macc1| Gene Symbol: Macc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163136.1 | NP_001156608.1 | 4 | 3015 | ||
| GenBank mRNA | AK028965.1 | - | 3 | 1337 | ||