siRNA Details
| Gene Symbol: Slc23a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017315.2 | NP_059011.1 | 7 | 760 | ||
| XM_006254602.2 | XP_006254664.1 | 9 | 1141 | |||
| GenBank mRNA | AF080452.1 | - | 7 | 760 | ||
| BC078851.1 | - | 7 | 760 | 7127602 | ||

Slc23a1
Genomic Map