siRNA Details
| Gene Symbol: Pappa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085376.2 | NP_001078845.1 | 22 | 6326 | ||
| GenBank mRNA | BC094560.1 | - | 12 | 2586 | ||
| CN700808.1 | - | 1 | 339 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pappa2| Gene Symbol: Pappa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085376.2 | NP_001078845.1 | 22 | 6326 | ||
| GenBank mRNA | BC094560.1 | - | 12 | 2586 | ||
| CN700808.1 | - | 1 | 339 | |||