siRNA Details
| Gene Symbol: Mylk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166030.1 | NP_001159502.1 | 12 | 5220 | ||
| GenBank mRNA | AK033351.1 | - | 1 | 1389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mylk4| Gene Symbol: Mylk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166030.1 | NP_001159502.1 | 12 | 5220 | ||
| GenBank mRNA | AK033351.1 | - | 1 | 1389 | ||