siRNA Details
| Gene Symbol: Spata31d1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083890.2 | NP_001077359.2 | 4 | 2193 | ||
| GenBank mRNA | AK133350.1 | - | 4 | 2194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata31d1c| Gene Symbol: Spata31d1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083890.2 | NP_001077359.2 | 4 | 2193 | ||
| GenBank mRNA | AK133350.1 | - | 4 | 2194 | ||