siRNA Details
Gene Symbol: Zfp72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081680.1 | NP_001075149.1 | 3 | 1417 | ||
GenBank mRNA | BC118036.1 | - | 3 | 1417 | 40106771 | |
BC118046.1 | - | 3 | 1417 | 40106772 |
