siRNA Details
| Gene Symbol: Ogdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081130.1 | NP_001074599.1 | 8 | 1063 | ||
| GenBank mRNA | BC157970.1 | - | 7 | 863 | 9007292 | |
| BC157971.1 | - | 7 | 863 | 9007293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ogdhl| Gene Symbol: Ogdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081130.1 | NP_001074599.1 | 8 | 1063 | ||
| GenBank mRNA | BC157970.1 | - | 7 | 863 | 9007292 | |
| BC157971.1 | - | 7 | 863 | 9007293 | ||