siRNA Details
| Gene Symbol: Setdb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081024.1 | NP_001074493.1 | 2 | 324 | ||
| GenBank mRNA | AK089197.1 | - | 2 | 324 | ||
| BC157926.1 | - | 2 | 194 | 9088099 | ||
| BC172021.1 | - | 2 | 194 | 9054307 | ||
| EU155090.1 | - | 2 | 186 | |||
| EU155091.1 | - | 2 | 186 | |||
| EU155092.1 | - | 2 | 204 | |||
| EU155093.1 | - | 2 | 186 | |||
| EU155094.1 | - | 2 | 186 | |||
| EU155097.1 | - | 3 | 245 | |||
| EU155098.1 | - | 2 | 204 | |||
| EU155099.1 | - | 2 | 204 | |||

Setdb2
Genomic Map