siRNA Details
| Gene Symbol: Card6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163138.1 | NP_001156610.1 | 4 | 1118 | ||
| GenBank mRNA | AK052796.1 | - | 3 | 1008 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Card6| Gene Symbol: Card6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163138.1 | NP_001156610.1 | 4 | 1118 | ||
| GenBank mRNA | AK052796.1 | - | 3 | 1008 | ||