siRNA Details
| Gene Symbol: Ttc38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033337.4 | NP_001028509.2 | 8, 9 | 871 | ||
| GenBank mRNA | AK078848.1 | - | 8, 9 | 832 | ||
| AK133636.1 | - | 7, 8 | 579 | |||
| AK200042.1 | - | 2, 3 | 123 | |||
| AK204138.1 | - | 2, 3 | 123 | |||
| BC024550.1 | - | 8, 9 | 821 | |||
| BC030849.1 | - | 8, 9 | 834 | |||
| BC132621.1 | - | 8, 9 | 856 | 40130898 | ||
| BC132623.1 | - | 8, 9 | 856 | 40130900 | ||

Ttc38
Genomic Map