siRNA Details
| Gene Symbol: Krtap24-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163141.1 | NP_001156613.1 | 1 | 748 | ||
| GenBank mRNA | AK037168.1 | - | 1 | 749 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap24-1| Gene Symbol: Krtap24-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163141.1 | NP_001156613.1 | 1 | 748 | ||
| GenBank mRNA | AK037168.1 | - | 1 | 749 | ||