siRNA Details
| Gene Symbol: Gm4951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033767.3 | NP_001028939.1 | 2 | 153 | ||
| GenBank mRNA | AK149583.1 | - | 2 | 151 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4951| Gene Symbol: Gm4951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033767.3 | NP_001028939.1 | 2 | 153 | ||
| GenBank mRNA | AK149583.1 | - | 2 | 151 | ||