siRNA Details
| Gene Symbol: Prf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017330.2 | NP_059026.2 | 2 | 540 | ||
| GenBank mRNA | BC089808.1 | - | 2 | 540 | 7378633 | |
| M33605.1 | - | 1 | 488 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prf1| Gene Symbol: Prf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017330.2 | NP_059026.2 | 2 | 540 | ||
| GenBank mRNA | BC089808.1 | - | 2 | 540 | 7378633 | |
| M33605.1 | - | 1 | 488 | |||