siRNA Details
| Gene Symbol: Mettl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143956.1 | NP_001137428.1 | 1 | 110 | ||
| GenBank mRNA | BC157906.1 | - | 1 | 110 | 9088108 | |
| BC158115.1 | - | 1 | 87 | 9088188 | ||
| BC172048.1 | - | 1 | 110 | 9054334 | ||
| BC172056.1 | - | 1 | 110 | |||

Mettl11b
Genomic Map