siRNA Details
| Gene Symbol: Dusp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033344.3 | NP_001028516.2 | 6 | 2904 | ||
| NM_001160049.1 | NP_001153521.1 | 6 | 2900 | |||
| GenBank mRNA | AK048603.1 | - | 6 | 2927 | ||
| AK142263.1 | - | 6 | 2905 | |||
| BC059034.1 | - | 2 | 2253 | |||
| BC117936.1 | - | 6 | 2881 | 40097789 | ||

Dusp27
Genomic Map