siRNA Details
| Gene Symbol: Zbtb34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085507.1 | NP_001078976.1 | 3 | 4705 | ||
| NM_001276332.1 | NP_001263261.1 | 2 | 4585 | |||
| GenBank mRNA | AK173321.1 | - | 2 | 4578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb34| Gene Symbol: Zbtb34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085507.1 | NP_001078976.1 | 3 | 4705 | ||
| NM_001276332.1 | NP_001263261.1 | 2 | 4585 | |||
| GenBank mRNA | AK173321.1 | - | 2 | 4578 | ||