siRNA Details
Gene Symbol: Hsd17b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017235.3 | NP_058931.1 | 8 | 948 | ||
XM_006250215.2 | XP_006250277.1 | 8 | 971 | |||
XM_006250216.2 | XP_006250278.1 | 8 | 971 | |||
XM_006250217.2 | XP_006250279.1 | 8 | 971 | |||
GenBank mRNA | FN803430.1 | - | 6 | 561 | ||
U44803.1 | - | 8 | 893 |
