siRNA Details
| Gene Symbol: D430041D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033347.2 | NP_001028519.2 | 21 | 7543 | ||
| GenBank mRNA | BC096034.1 | - | 1 | 1125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D430041D05Rik| Gene Symbol: D430041D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033347.2 | NP_001028519.2 | 21 | 7543 | ||
| GenBank mRNA | BC096034.1 | - | 1 | 1125 | ||