siRNA Details
Gene Symbol: D430041D05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033347.2 | NP_001028519.2 | 21 | 5494 | ||
GenBank mRNA | AK052518.1 | - | 12 | 3397 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D430041D05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033347.2 | NP_001028519.2 | 21 | 5494 | ||
GenBank mRNA | AK052518.1 | - | 12 | 3397 |