siRNA Details
| Gene Symbol: D430041D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033347.2 | NP_001028519.2 | 21 | 9168 | ||
| GenBank mRNA | AK138032.1 | - | 1 | 2368 | ||
| AK165696.1 | - | 1 | 339 | |||
| BC096034.1 | - | 1 | 2750 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D430041D05Rik| Gene Symbol: D430041D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033347.2 | NP_001028519.2 | 21 | 9168 | ||
| GenBank mRNA | AK138032.1 | - | 1 | 2368 | ||
| AK165696.1 | - | 1 | 339 | |||
| BC096034.1 | - | 1 | 2750 | |||