siRNA Details
| Gene Symbol: Wdr76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290986.1 | NP_001277915.1 | 2 | 335 | ||
| NM_001290987.1 | NP_001277916.1 | 2 | 350 | |||
| NM_030234.2 | NP_084510.2 | 2 | 350 | |||
| GenBank mRNA | AK020004.1 | - | 2 | 303 | ||
| AK031592.1 | - | 2 | 276 | |||
| AK051037.1 | - | 2 | 319 | |||
| AK162181.1 | - | 2 | 301 | |||
| BC117821.1 | - | 2 | 289 | |||
| BC117822.1 | - | 2 | 289 | 40092839 | ||
| BY720216.1 | - | 2 | 350 | |||

Wdr76
Genomic Map