siRNA Details
| Gene Symbol: Atp8b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080944.3 | NP_001074413.2 | 17 | 1775 | ||
| GenBank mRNA | AK086607.1 | - | 16 | 1763 | ||
| AK155448.1 | - | 17 | 1785 | |||
| AK169967.1 | - | 17 | 1798 | |||
| AK173307.1 | - | 17 | 1775 | |||

Atp8b4
Genomic Map