siRNA Details
| Gene Symbol: Kcng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081134.1 | NP_001074603.1 | 2 | 1146 | ||
| GenBank mRNA | BC147251.1 | - | 2 | 1078 | 9056302 | |
| BC147252.1 | - | 2 | 1078 | 9056297 | ||
| BC147715.1 | - | 2 | 1078 | 9087474 | ||

Kcng1
Genomic Map