siRNA Details
| Gene Symbol: D3Ertd254e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101478.1 | NP_001094948.1 | 4 | 972 | ||
| GenBank mRNA | AK153849.1 | - | 4 | 922 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D3Ertd254e| Gene Symbol: D3Ertd254e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101478.1 | NP_001094948.1 | 4 | 972 | ||
| GenBank mRNA | AK153849.1 | - | 4 | 922 | ||