siRNA Details
| Gene Symbol: Slc44a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081263.1 | NP_001074732.1 | 22 | 2578 | ||
| GenBank mRNA | BC068154.1 | - | 9 | 3672 | ||
| BC086641.1 | - | 22 | 2578 | 6417957 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc44a5| Gene Symbol: Slc44a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081263.1 | NP_001074732.1 | 22 | 2578 | ||
| GenBank mRNA | BC068154.1 | - | 9 | 3672 | ||
| BC086641.1 | - | 22 | 2578 | 6417957 | ||