siRNA Details
| Gene Symbol: Lrrc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081358.2 | NP_001074827.1 | 25 | 5599 | ||
| NM_001291452.1 | NP_001278381.1 | 24 | 5458 | |||
| NM_001291453.1 | NP_001278382.1 | 23 | 5338 | |||
| GenBank mRNA | AK122498.1 | - | 24 | 4593 | ||
| BC138450.1 | - | 23 | 5245 | 8861470 | ||

Lrrc7
Genomic Map