siRNA Details
| Gene Symbol: Lrrc38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162983.1 | NP_001156455.1 | 1 | 64 | ||
| GenBank mRNA | AK038658.1 | - | 1 | 26 | ||
| AK078190.1 | - | 1 | 38 | |||
| BC129963.1 | - | 1 | 57 | |||
| BC129964.1 | - | 1 | 64 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc38| Gene Symbol: Lrrc38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162983.1 | NP_001156455.1 | 1 | 64 | ||
| GenBank mRNA | AK038658.1 | - | 1 | 26 | ||
| AK078190.1 | - | 1 | 38 | |||
| BC129963.1 | - | 1 | 57 | |||
| BC129964.1 | - | 1 | 64 | |||