siRNA Details
| Gene Symbol: Klhl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033352.3 | NP_001028524.1 | 4 | 3655 | ||
| GenBank mRNA | AK007786.1 | - | 1 | 1482 | ||
| AK052203.1 | - | 4 | 3628 | |||
| AK154493.1 | - | 4 | 3656 | |||
| AK158141.1 | - | 5 | 3615 | |||
| BC060201.1 | - | 4 | 3644 | |||

Klhl21
Genomic Map