siRNA Details
| Gene Symbol: Klhl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033352.3 | NP_001028524.1 | 3, 4 | 1551 | ||
| GenBank mRNA | AK052203.1 | - | 3, 4 | 1526 | ||
| AK154493.1 | - | 3, 4 | 1552 | |||
| AK158141.1 | - | 3, 4 | 1527 | |||
| BC060201.1 | - | 3, 4 | 1540 | |||
| BC141158.1 | - | 3, 4 | 1525 | 9055770 | ||
| BC141159.1 | - | 3, 4 | 1525 | 9055771 | ||

Klhl21
Genomic Map