siRNA Details
| Gene Symbol: Prickle2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081146.2 | NP_001074615.1 | 9 | 3262 | ||
| NM_001134459.1 | NP_001127931.1 | 8 | 3341 | |||
| NM_001134460.2 | NP_001127932.1 | 8 | 3663 | |||
| NM_001134461.1 | NP_001127933.1 | 8 | 3158 | |||
| GenBank mRNA | AK032631.1 | - | 8 | 3670 | ||
| AK147576.1 | - | 8 | 3341 | |||
| AK147628.1 | - | 8 | 3341 | |||
| BC099897.1 | - | 9 | 3233 | 30621797 | ||

Prickle2
Genomic Map