siRNA Details
| Gene Symbol: Gltscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081418.1 | NP_001074887.1 | 14 | 5122 | ||
| GenBank mRNA | AK220278.1 | - | 8 | 5795 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gltscr1| Gene Symbol: Gltscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081418.1 | NP_001074887.1 | 14 | 5122 | ||
| GenBank mRNA | AK220278.1 | - | 8 | 5795 | ||