siRNA Details
| Gene Symbol: Bcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017253.2 | NP_058949.1 | 7 | 932 | ||
| XM_008763370.1 | XP_008761592.1 | 7 | 945 | |||
| XM_008763371.1 | XP_008761593.1 | 7 | 1195 | |||
| XM_008763372.1 | XP_008761594.1 | 8 | 1021 | |||
| XM_008763373.1 | XP_008761595.1 | 7 | 1124 | |||
| GenBank mRNA | AF165887.1 | - | 5 | 689 | ||
| AJ278701.1 | - | 7 | 926 | |||
| BC087710.1 | - | 7 | 932 | 7315507 | ||
| U35774.1 | - | 7 | 898 | |||

Bcat1
Genomic Map