siRNA Details
| Gene Symbol: Usp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177412.1 | NP_001170883.1 | 2, 3 | 805 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp35| Gene Symbol: Usp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177412.1 | NP_001170883.1 | 2, 3 | 805 | ||