siRNA Details
| Gene Symbol: Myo16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081397.1 | NP_001074866.1 | 24 | 3149 | ||
| GenBank mRNA | AB429291.1 | - | 21 | 2655 | ||
| AK034257.1 | - | 1 | 23 | |||
| AK082350.1 | - | 24 | 2885 | |||
| AK220356.1 | - | 5 | 581 | |||
| BC082311.1 | - | 24 | 3015 | |||
| BC151049.1 | - | 24 | 3103 | 9087962 | ||
| BC151051.1 | - | 24 | 3104 | 9087964 | ||
| BC157966.1 | - | 24 | 3104 | 9007290 | ||
| BC172122.1 | - | 24 | 3104 | 9054408 | ||

Myo16
Genomic Map