siRNA Details
Gene Symbol: Cntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170787.1 | NP_001164258.1 | 6 | 917 | ||
GenBank mRNA | AK032746.1 | - | 2 | 685 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170787.1 | NP_001164258.1 | 6 | 917 | ||
GenBank mRNA | AK032746.1 | - | 2 | 685 |