siRNA Details
| Gene Symbol: C2cd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163143.1 | NP_001156615.1 | 2 | 1305 | ||
| GenBank mRNA | AK080627.1 | - | 1 | 845 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2cd4a| Gene Symbol: C2cd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163143.1 | NP_001156615.1 | 2 | 1305 | ||
| GenBank mRNA | AK080627.1 | - | 1 | 845 | ||