siRNA Details
| Gene Symbol: Slc35g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101483.1 | NP_001094953.1 | 2 | 1385 | ||
| GenBank mRNA | AV173252.2 | - | 1 | 436 | ||
| CK794116.1 | - | 1 | 312 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35g2| Gene Symbol: Slc35g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101483.1 | NP_001094953.1 | 2 | 1385 | ||
| GenBank mRNA | AV173252.2 | - | 1 | 436 | ||
| CK794116.1 | - | 1 | 312 | |||