siRNA Details
Gene Symbol: Gm4981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034869.2 | NP_001030041.1 | 1 | 1601 | ||
GenBank mRNA | AK145460.1 | - | 1 | 1602 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034869.2 | NP_001030041.1 | 1 | 1601 | ||
GenBank mRNA | AK145460.1 | - | 1 | 1602 |