siRNA Details
Gene Symbol: Gm364 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128625.2 | NP_001122097.1 | 1, 2 | 221 | ||
GenBank mRNA | AK030183.1 | - | 217 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm364 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128625.2 | NP_001122097.1 | 1, 2 | 221 | ||
GenBank mRNA | AK030183.1 | - | 217 |