siRNA Details
| Gene Symbol: Gm364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128625.2 | NP_001122097.1 | 5, 6 | 641 | ||
| GenBank mRNA | AK030183.1 | - | 637 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm364| Gene Symbol: Gm364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128625.2 | NP_001122097.1 | 5, 6 | 641 | ||
| GenBank mRNA | AK030183.1 | - | 637 | |||