siRNA Details
| Gene Symbol: Gm614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033362.2 | NP_001028534.1 | 2 | 220 | ||
| GenBank mRNA | AK131750.1 | - | 2 | 221 | ||
| BC048573.1 | - | 2 | 230 | |||
| BC147523.1 | - | 2 | 217 | 9056515 | ||
| BC147537.1 | - | 2 | 217 | 9056528 | ||

Gm614
Genomic Map