siRNA Details
| Gene Symbol: Timm44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017267.1 | NP_058963.1 | 3 | 328 | ||
| GenBank mRNA | AB006452.1 | - | 3 | 328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timm44| Gene Symbol: Timm44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017267.1 | NP_058963.1 | 3 | 328 | ||
| GenBank mRNA | AB006452.1 | - | 3 | 328 | ||