siRNA Details
| Gene Symbol: Olfr193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011791.1 | NP_001011791.1 | 1 | 479 | ||
| GenBank mRNA | BC147133.1 | - | 1 | 504 | 9056183 | |
| BC147134.1 | - | 1 | 502 | 9056182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr193| Gene Symbol: Olfr193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011791.1 | NP_001011791.1 | 1 | 479 | ||
| GenBank mRNA | BC147133.1 | - | 1 | 504 | 9056183 | |
| BC147134.1 | - | 1 | 502 | 9056182 | ||