siRNA Details
| Gene Symbol: Olfr598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011793.1 | NP_001011793.1 | 1 | 228 | ||
| Gene Symbol: Olfr597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC150974.1 | - | 1 | 243 | 9087887 | |

Olfr598
Genomic Map