siRNA Details
| Gene Symbol: Gpam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017274.1 | NP_058970.1 | 15 | 1819 | ||
| XM_006231625.2 | XP_006231687.1 | 15 | 2210 | |||
| XM_006231626.2 | XP_006231688.1 | 16 | 2091 | |||
| XM_008760534.1 | XP_008758756.1 | 15 | 1761 | |||
| GenBank mRNA | AF021348.1 | - | 15 | 1819 | ||
| U36771.2 | - | 14 | 1595 | |||

Gpam
Genomic Map