siRNA Details
| Gene Symbol: Olfr597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011845.2 | NP_001011845.2 | 1 | 909 | ||
| GenBank mRNA | BC150974.1 | - | 1 | 936 | 9087887 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr597| Gene Symbol: Olfr597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011845.2 | NP_001011845.2 | 1 | 909 | ||
| GenBank mRNA | BC150974.1 | - | 1 | 936 | 9087887 | |