siRNA Details
| Gene Symbol: Olfr391-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159775.1 | NP_001153247.1 | 2 | 427 | ||
| GenBank mRNA | CB173237.1 | - | 2 | 413 | ||
| CB174118.1 | - | 1 | 218 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr391-ps| Gene Symbol: Olfr391-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159775.1 | NP_001153247.1 | 2 | 427 | ||
| GenBank mRNA | CB173237.1 | - | 2 | 413 | ||
| CB174118.1 | - | 1 | 218 | |||