siRNA Details
| Gene Symbol: Olfr1274-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146263.2 | NP_666375.2 | 1 | 984 | ||
| GenBank mRNA | BC145937.1 | - | 984 | 40131240 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1274-ps| Gene Symbol: Olfr1274-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146263.2 | NP_666375.2 | 1 | 984 | ||
| GenBank mRNA | BC145937.1 | - | 984 | 40131240 | ||