siRNA Details
| Gene Symbol: Olfr291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146415.2 | NP_666527.1 | 2 | 130 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB172982.1 | - | 114 | Chr. - - [-] on Build | ||

Olfr291
Genomic Map