siRNA Details
| Gene Symbol: Olfr291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146415.2 | NP_666527.1 | 3 | 854 | ||
| GenBank mRNA | BC100693.1 | - | 1 | 648 | ||
| BC100694.1 | - | 1 | 647 | |||
| BC100695.1 | - | 1 | 648 | |||
| BC114974.1 | - | 1 | 648 | 40043317 | ||
| BC115740.1 | - | 1 | 648 | |||

Olfr291
Genomic Map